Learn More
Medchemexpress LLC DODMA | 104162-47-2 | 99.6% | 620.09 | 10 MM 1 ML
SDP
Catalog No. 5000436944
Encompass_Preferred
DODMA is a cationic lipid used to prepare lipid nanoparticles and for drug delivery. It features a pH-titrable head group that is neutral at physiological pH for systemic circulation, and protonates in the acidic environment of endosomes, promoting fusion with the endosome membrane and aiding nucleic acid release.
  • Cationic lipid
  • Can be used to prepare lipid nanoparticles
  • Can be used for drug delivery
  • pH-titrable head group, neutral at physiological pH
  • Promotes fusion with endosome membrane and nucleic acid release in acidic endosomal environments
Catalog No. 50-004-36944 Supplier Medchemexpress LLC Supplier No. HY11275510MM/1ML
DODMA is a cationic lipid used to prepare lipid nanoparticles and for drug delivery. It features a pH-titrable head group that is neutral at physiological pH for systemic circulation, and protonates in the acidic environment of endosomes, promoting fusion with the endosome membrane and aiding nucleic acid release.
  • Cationic lipid
  • Can be used to prepare lipid nanoparticles
  • Can be used for drug delivery
  • pH-titrable head group, neutral at physiological pH
  • Promotes fusion with endosome membrane and nucleic acid release in acidic endosomal environments