Learn More
Medchemexpress LLC DODMA | 104162-47-2 | 99.6% | 620.09 | 100 MG
SDP
Catalog No. 5000437396
Encompass_Preferred
DODMA is a cationic lipid used to prepare lipid nanoparticles for drug delivery. Its unique pH-titratable head group maintains neutrality at physiological pH, aiding systemic circulation. Upon encountering the acidic environment of endosomes, the head group protonates, facilitating membrane fusion and promoting nucleic acid release.
  • Cationic lipid for lipid nanoparticle preparation
  • Used in drug delivery applications
  • pH-titratable head group for systemic circulation
  • Promotes fusion with endosome membrane
  • Facilitates nucleic acid release
Catalog No. 50-004-37396 Supplier Medchemexpress LLC Supplier No. HY112755100MG
DODMA is a cationic lipid used to prepare lipid nanoparticles for drug delivery. Its unique pH-titratable head group maintains neutrality at physiological pH, aiding systemic circulation. Upon encountering the acidic environment of endosomes, the head group protonates, facilitating membrane fusion and promoting nucleic acid release.
  • Cationic lipid for lipid nanoparticle preparation
  • Used in drug delivery applications
  • pH-titratable head group for systemic circulation
  • Promotes fusion with endosome membrane
  • Facilitates nucleic acid release