Learn More
Medchemexpress LLC Dodma | 104162-47-2 | 99.60% | 620.09 | 50 MG
SDP
Catalog No. 5000437702
Encompass_Preferred
DODMA is a cationic lipid used in the preparation of lipid nanoparticles for drug delivery. Its pH-titratable head group remains neutral at physiological pH for systemic circulation, but protonates in the acidic environment of endosomes, facilitating fusion with the endosome membrane and aiding nucleic acid release.
  • Cationic lipid
  • Can be used to prepare lipid nanoparticles
  • Can be used for drug delivery
Catalog No. 50-004-37702 Supplier Medchemexpress LLC Supplier No. HY11275550MG
DODMA is a cationic lipid used in the preparation of lipid nanoparticles for drug delivery. Its pH-titratable head group remains neutral at physiological pH for systemic circulation, but protonates in the acidic environment of endosomes, facilitating fusion with the endosome membrane and aiding nucleic acid release.
  • Cationic lipid
  • Can be used to prepare lipid nanoparticles
  • Can be used for drug delivery