Learn More
Medchemexpress LLC Dodma | 104162-47-2 | 99.60% | 620.09 | 500 MG
SDP
Catalog No. 5000436946
Encompass_Preferred
DODMA is a cationic lipid that can be utilized in the preparation of lipid nanoparticles. This compound is suitable for drug delivery applications. It features a pH-titrable head group, which remains neutral at physiological pH, aiding systemic circulation. In the acidic conditions of endosomes, the head group protonates, promoting fusion with the endosome membrane and facilitating nucleic acid release.
  • Can be used to prepare lipid nanoparticles.
  • Applicable for drug delivery.
  • Possesses a pH-titrable head group that is neutral at physiological pH for systemic circulation.
  • Head group protonates in acidic endosomal environments, enhancing fusion with the endosome membrane and nucleic acid release.
Catalog No. 50-004-36946 Supplier Medchemexpress LLC Supplier No. HY112755500MG
DODMA is a cationic lipid that can be utilized in the preparation of lipid nanoparticles. This compound is suitable for drug delivery applications. It features a pH-titrable head group, which remains neutral at physiological pH, aiding systemic circulation. In the acidic conditions of endosomes, the head group protonates, promoting fusion with the endosome membrane and facilitating nucleic acid release.
  • Can be used to prepare lipid nanoparticles.
  • Applicable for drug delivery.
  • Possesses a pH-titrable head group that is neutral at physiological pH for systemic circulation.
  • Head group protonates in acidic endosomal environments, enhancing fusion with the endosome membrane and nucleic acid release.