Learn More
Kyfora Bio DODMA 500 MG
Catalog No. 502559841
Encompass_Preferred
DODMA (1,2-dioleyloxy-3-dimethylaminopropane) is an ionizable lipid with a protonatable tertiary amine head group that has a positive charge at low pH. DODMA is highly efficient at encapsulating cargo and forming lipoplexes, which readily pass through the cell membrane. These properties make DODMA highly suitable for nucleic acid and drug delivery applications.
Catalog No. 50-255-9841 Supplier Kyfora Bio Supplier No. 14478500
DODMA (1,2-dioleyloxy-3-dimethylaminopropane) is an ionizable lipid with a protonatable tertiary amine head group that has a positive charge at low pH. DODMA is highly efficient at encapsulating cargo and forming lipoplexes, which readily pass through the cell membrane. These properties make DODMA highly suitable for nucleic acid and drug delivery applications.