Learn More
Medchemexpress LLC Dofequidar (fumarate) | 153653-30-6 | 98.0% | 597.66 | 5 MG
SDP
Catalog No. 5000385413
Encompass_Preferred
Dofequidar fumarate (MS-209) is an orally active quinoline compound that blocks P-glycoprotein (P-gp) and multidrug resistance-associated protein-1 (MDR-1). It has a highly potent reversing effect on multidrug-resistant tumor cells. Dofequidar fumarate competitively inhibits ABCB1/P-gp and ABCC1/MRP-1, blocking the efflux of chemotherapeutic agents, increasing drug concentration in cancer cells, and enhancing the chemotherapeutic effect.
  • Orally active
  • Blocks P-glycoprotein (P-gp) and multidrug resistance-associated protein-1 (MDR-1)
  • Highly potent reversing effect on multidrug-resistant tumor cells
  • Competitively inhibits ABCB1/P-gp and ABCC1/MRP-1
  • Blocks the efflux of chemotherapeutic agents
  • Increases drug concentration in cancer cells
  • Enhances chemotherapeutic effect
Catalog No. 50-003-85413 Supplier Medchemexpress LLC Supplier No. HY17013A5MG
Add to Cart
Add to Cart
Dofequidar fumarate (MS-209) is an orally active quinoline compound that blocks P-glycoprotein (P-gp) and multidrug resistance-associated protein-1 (MDR-1). It has a highly potent reversing effect on multidrug-resistant tumor cells. Dofequidar fumarate competitively inhibits ABCB1/P-gp and ABCC1/MRP-1, blocking the efflux of chemotherapeutic agents, increasing drug concentration in cancer cells, and enhancing the chemotherapeutic effect.
  • Orally active
  • Blocks P-glycoprotein (P-gp) and multidrug resistance-associated protein-1 (MDR-1)
  • Highly potent reversing effect on multidrug-resistant tumor cells
  • Competitively inhibits ABCB1/P-gp and ABCC1/MRP-1
  • Blocks the efflux of chemotherapeutic agents
  • Increases drug concentration in cancer cells
  • Enhances chemotherapeutic effect