Learn More
Apexbio Technology LLC Dofetilide 115256-11-6 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061930
Encompass_Preferred
Dofetilide (CAS 115256-11-6) chemically designated as UK 68789 is a class III antiarrhythmic small molecule that selectively inhibits the rapid delayed rectifier potassium channel (IKr) This inhibition leads to prolonged ventricular action potential duration and extended effective refractory periods in cardiac cells Noted for favorable oral bioavailability Dofetilide is extensively utilized in cardiac electrophysiological research including studies aimed at elucidating arrhythmogenic mechanisms linked to IKr channel function assessing novel antiarrhythmic candidates and exploring physiological and pathological aspects of cardiac rhythm regulation
Catalog No. 50-000-61930 Supplier Apexbio Technology LLC Supplier No. A84175.1
Dofetilide (CAS 115256-11-6) chemically designated as UK 68789 is a class III antiarrhythmic small molecule that selectively inhibits the rapid delayed rectifier potassium channel (IKr) This inhibition leads to prolonged ventricular action potential duration and extended effective refractory periods in cardiac cells Noted for favorable oral bioavailability Dofetilide is extensively utilized in cardiac electrophysiological research including studies aimed at elucidating arrhythmogenic mechanisms linked to IKr channel function assessing novel antiarrhythmic candidates and exploring physiological and pathological aspects of cardiac rhythm regulation