Learn More
Selleck Chemical LLC Dofetilide S1658-200mg
SDP
Supplier:  Selleck Chemical LLC S1658200MG
Encompass_Preferred
Dofetilide (UK-68798) is a selective potassium channel ((hERG)) blocker used as a Class III antiarrhythmic drug
Catalog No. 50-310-8468