Learn More
Cayman Chemical DofetIlIde-d4 1mg
SDP
Catalog No. 502934356
Encompass_Preferred
An internal standard for the quantification of dofetilide by GC- or LC-MS
Catalog No. 50-293-4356 Supplier Cayman Chemical Supplier No. 264511 MG
Add to Cart
Add to Cart
An internal standard for the quantification of dofetilide by GC- or LC-MS