Learn More
Cayman Chemical DofetIlIde-d4 500ug
SDP
Catalog No. 502933613
Encompass_Preferred
An internal standard for the quantification of dofetilide by GC- or LC-MS
Catalog No. 50-293-3613 Supplier Cayman Chemical Supplier No. 26451500 UG
Add to Cart
Add to Cart
An internal standard for the quantification of dofetilide by GC- or LC-MS