Learn More
World Precision Instrument Dog Adaptor
SDP
Catalog No. 502539108
Encompass
The Dog/Monkey Adaptor is available only for the Large Animal Stereotaxic, 4 Manipulator Arms (505326). To fit any size dog, it should be used with 505341 and 505342 stereotaxic instruments, which have two sets of ear bar holders at different spacings from the front, to accommodate a wide size range of dogs.
Catalog No. 50-253-9108 Supplier World Precision Instrument Supplier No. 505343
The Dog/Monkey Adaptor is available only for the Large Animal Stereotaxic, 4 Manipulator Arms (505326). To fit any size dog, it should be used with 505341 and 505342 stereotaxic instruments, which have two sets of ear bar holders at different spacings from the front, to accommodate a wide size range of dogs.