Learn More
Biomatik Corporation Dog Apolipoprotein A1 (APOA1) ELISA Kit
Supplier:  Biomatik Corporation EKU02479
Encompass_Preferred
Dog Apolipoprotein A1 (APOA1) ELISA Kit for detecting APOA1 in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.625-40ng/ml. Sensitivity: 0.267ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02648. Target Synonyms: Apo-A1; ApoA-1 Milano; ProapoA-I; Proapolipoprotein A-I; Truncated apolipoprotein A-I. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6330