Learn More
Biomatik Corporation Dog Apolipoprotein E (APOE) ELISA Kit
Supplier:  Biomatik Corporation EKU02516
Encompass_Preferred
Dog Apolipoprotein E (APOE) ELISA Kit for detecting APOE in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 1.56-100ng/ml. Sensitivity: 0.65ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18649. Gene ID: 476438. Target Synonyms: Apo-E; AD2; Apoprotein; Alzheimer Disease 2 (E4-Associated,Late Onset. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9125