Learn More
Biomatik Corporation Dog Bone Morphogenetic Protein 2 (BMP2) ELISA Kit
Supplier:  Biomatik Corporation EKU11650
Encompass_Preferred
Dog Bone Morphogenetic Protein 2 (BMP2) ELISA Kit for detecting BMP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: J9P401. Target Synonyms: BMP2A; BMP-2A; Hemochromatosis Modifier. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4957