Learn More
Biomatik Corporation Dog Bone Morphogenetic Protein 4 (BMP4) CLIA Kit
Supplier:  Biomatik Corporation EKU09937
Encompass_Preferred
Dog Bone Morphogenetic Protein 4 (BMP4) CLIA Kit for detecting BMP4 in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.16pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: F1PDL1. Target Synonyms: BMP2B; BMP2-B; BMP2B1; ZYME; DVR4; Bone morphogenetic protein 2B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8399