Learn More
American Research Products Inc Dog C C motif chemokine 5 CCL
Catalog No. 5000327612
Encompass
Dog C-C motif chemokine 5 (CCL5) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 081 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27612 Supplier American Research Products Inc Supplier No. CSBEL004800DO
Dog C-C motif chemokine 5 (CCL5) ELISA kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 081 ng/mL Sample serum plasma tissue homogenates Method Sandwich