Learn More
Innovative Research Inc Dog C-ReAct Prtn Kit
Supplier:  Innovative Research Inc ICNCRPKT
Encompass_Preferred
Canine C-Reactive Protein ELISA Kit - This Kit is a sensitive two-site enzyme linked ELISA for measuring Canine C-Reactive Protein (CRP) in biological samples. Strip well format for up to 96 tests. In this assay the CRP present in samples reacts with the anti-CRP antibodies which have been adsorbed to the surface of polystyrene microtitre wells. After the removal of unbound proteins by washing, anti-CRP antibodies conjugated with horseradish peroxidase (HRP), are added. These enzyme-labeled antibodies form complexes with the previously bound CRP. Following washings, the enzyme bound to the immunosorbent is assayed by the addition of a chromogenic substrate, 3,3AE,5,5AE-tetramethylbenzidine (TMB). The bound enzyme varies directly with the concentration of CRP in the sample; thus, the absorbance, at 450 nm, is a measure of the concentration of CRP in the sample. The quantity of CRP in the test sample can be interpolated from the standard curve constructed from the standards
Catalog No. 50-274-4609