Learn More
Biomatik Corporation Dog Calprotectin (CALPRO) CLIA Kit
Supplier:  Biomatik Corporation EKU12421
Encompass_Preferred
Dog Calprotectin (CALPRO) CLIA Kit for detecting CALPRO in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.14-100ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: C0LQL0. Gene ID: 490461. Target Synonyms: MRP-8/MRP-14; S100A8/A9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7168