Learn More
American Research Products Inc Dog circulating immune compleX
Catalog No. 5000326411
Encompass
Dog circulating immune compleX (CIC) ELISA kit from CUSABIO Detection Range 4 68 ng/ml - 1200 ng/ml Sensitivity 4 68 ng/ml Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26411 Supplier American Research Products Inc Supplier No. CSBEQ027971DO
Dog circulating immune compleX (CIC) ELISA kit from CUSABIO Detection Range 4 68 ng/ml - 1200 ng/ml Sensitivity 4 68 ng/ml Sample serum plasma tissue homogenates Method Competitive