Learn More
Biomatik Corporation Dog Clusterin, Clu Elisa Kit
Supplier:  Biomatik Corporation EKC31055
Encompass_Preferred
Dog Clusterin, Clu Elisa Kit for detecting Clusterin in serum, plasma, tissue homogenates. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 7.813-500ng/ml. Sensitivity: 4.963ng/ml. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Accession Number: P25473; CLU. Target Synonyms: CLUClusterin; Glycoprotein 80; Gp80) [Cleaved into: Clusterin beta chain; Clusterin alpha chain]. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4747