Learn More
ACROBiosystems Dog Complement C2 25ug
Supplier:  ACROBiosystems CO2D82E425UG
Encompass_Preferred
Biotinylated Dog Complement C2 Protein HisAvitag (CO2-D82E4) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Leu 784 (Accession # A0A8I3NQQ5).
Catalog No. 50-253-5230