Learn More
Life Diagnostics Inc DOG CPR LYOPHILIZED 1MG
SDP
Catalog No. NC3807127
Encompass_Preferred
CRP is a positive acute phase reactant present in blood that is expressed in the liver. It is used as a biomarker for cardiovascular and inflammatory disease. It is a pentameric protein comprised of five identical 25 kDa polypeptide chains. On SDS PAGE gels two bands are apparent. This is due to differential glycosylation of the polypeptide chains. During the acute phase reaction CRP levels increase several 100-fold in dogs.
Catalog No. NC3807127 Supplier Life Diagnostics Inc Supplier No. 8101L
CRP is a positive acute phase reactant present in blood that is expressed in the liver. It is used as a biomarker for cardiovascular and inflammatory disease. It is a pentameric protein comprised of five identical 25 kDa polypeptide chains. On SDS PAGE gels two bands are apparent. This is due to differential glycosylation of the polypeptide chains. During the acute phase reaction CRP levels increase several 100-fold in dogs.