Learn More
American Research Products Inc Dog Erythropoietin EPO ELISA
Catalog No. 5000326077
Encompass
Dog Erythropoietin EPO ELISA Kit from CUSABIO Detection Range 1 56 mIU/mL-100 mIU/mL Sensitivity 0 39 mIU/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26077 Supplier American Research Products Inc Supplier No. CSBE11303C
Dog Erythropoietin EPO ELISA Kit from CUSABIO Detection Range 1 56 mIU/mL-100 mIU/mL Sensitivity 0 39 mIU/mL Sample serum plasma tissue homogenates Method Sandwich