Learn More
Cayman Chemical Dog Farnesoid X Receptor Reporter Assay System, 96 Well
SDP
Catalog No. 502946997
Encompass_Preferred
Dog FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. D00601-32, D00601]
Catalog No. 50-294-6997 Supplier Cayman Chemical Supplier No. 3350796 WELL
Dog FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. D00601-32, D00601]