Learn More
Cayman Chemical Dog Farnesoid X Receptor Reporter Assay System, 3x32 Well
SDP
Catalog No. 502947521
Encompass_Preferred
Dog FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. D00601-32, D00601]
Catalog No. 50-294-7521 Supplier Cayman Chemical Supplier No. 335073X32 WELL
Dog FXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. D00601-32, D00601]