Learn More
Biomatik Corporation Dog Ghrelin (GHRL) ELISA Kit
Supplier:  Biomatik Corporation EKU12388
Encompass_Preferred
Dog Ghrelin (GHRL) ELISA Kit for detecting GHRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.27pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9BEF8. Gene ID: 403587. Target Synonyms: MTLRP; Growth Hormone-Releasing Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4479