Learn More
Biomatik Corporation Dog Hepatocyte Growth Factor (HGF) CLIA Kit
Supplier:  Biomatik Corporation EKU09739
Encompass_Preferred
Dog Hepatocyte Growth Factor (HGF) CLIA Kit for detecting HGF in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.53pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q867B7. Gene ID: 403441. Target Synonyms: HGF; F-TCF; HGFB; HPTA; SF; Scatter Factor; Hepapoietin A; Fibroblast-Derived Tumor Cytotoxic Factor; Lung Fibroblast-Derived Mitogen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8449