Learn More
Immunology Consultants Dog IgG ELISA Kit
SDP
Supplier:  Immunology Consultants E40G
Encompass_Preferred
The Dog IgG test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgG in dog biological samples.
Catalog No. 50-240-6535