Learn More
Rockland Immunochemicals Dog IgG (H&L) Antibody 2 mL
SDP
Catalog No. 501943781
Encompass_Preferred
Secreted as part of the adaptive immune response by plasma B cells immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses bacteria as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them) activation of the compliment cascade and opsinization for phagocytosis. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition. Anti-Dog IgG (H&L) antibody is ideal for investigators in Immunology Cancer and Microbiology research.
Catalog No. 50-194-3781 Supplier Rockland Immunochemicals Supplier No. 1044102
Secreted as part of the adaptive immune response by plasma B cells immunoglobulin G constitutes 75% of serum immunoglobulins. Immunoglobulin G binds to viruses bacteria as well as fungi and facilitates their destruction or neutralization via agglutination (and thereby immobilizing them) activation of the compliment cascade and opsinization for phagocytosis. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition. Anti-Dog IgG (H&L) antibody is ideal for investigators in Immunology Cancer and Microbiology research.