Learn More
Rockland Immunochemicals Dog IgG Lyophilized 5mg
SDP
Catalog No. NC3038160
Encompass_Preferred
Dog IgG LyophilizedItem No.004-0102-0005Size: 5 mgPhysical State: LyophilizedApplications: SDS-PAGE, Biochemical Assay, Functional AssayOrigin: Dog
Catalog No. NC3038160 Supplier Rockland Immunochemicals Supplier No. 00401020005
Dog IgG LyophilizedItem No.004-0102-0005Size: 5 mgPhysical State: LyophilizedApplications: SDS-PAGE, Biochemical Assay, Functional AssayOrigin: Dog