Learn More
Biomatik Corporation Dog Insulin (INS) ELISA Kit
Supplier:  Biomatik Corporation EKU10142
Encompass_Preferred
Dog Insulin (INS) ELISA Kit for detecting INS in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 61.7-5,000pg/ml. Sensitivity: 23.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01321. Gene ID: 483665. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4468