Learn More
American Research Products Inc Dog insulin like growth factor
Supplier:  American Research Products Inc CSBEL011097DO
Encompass
Dog insulin-like growth factor binding protein 3 (IGFBP3) ELISA kit from CUSABIO Detection Range 3 12 ng/mL-200 ng/mL Sensitivity 0 78 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27854