Learn More
American Research Products Inc Dog insulin like growth factor
Catalog No. 5000327233
Encompass
Dog insulin-like growth factor 1 (IGF1) ELISA kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 8 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27233 Supplier American Research Products Inc Supplier No. CSBEL011086DO
Dog insulin-like growth factor 1 (IGF1) ELISA kit from CUSABIO Detection Range 31 25 ng/mL-2000 ng/mL Sensitivity 7 8 ng/mL Sample serum plasma tissue homogenates Method Sandwich