Learn More
American Research Products Inc Dog intercellular adhesion mol
Catalog No. 5000326731
Encompass
Dog intercellular adhesion molecule 1 ICAM-1/CD54 ELISA Kit from CUSABIO Detection Range 25 ng/mL-1600 ng/mL Sensitivity 6 25 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26731 Supplier American Research Products Inc Supplier No. CSBE08898C
Dog intercellular adhesion molecule 1 ICAM-1/CD54 ELISA Kit from CUSABIO Detection Range 25 ng/mL-1600 ng/mL Sensitivity 6 25 ng/mL Sample serum plasma tissue homogenates Method Sandwich