Learn More
American Research Products Inc Dog Interleukin 1 receptor ant
Catalog No. 5000327425
Encompass
Dog Interleukin-1 receptor antagonist protein (IL1RN) ELISA kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27425 Supplier American Research Products Inc Supplier No. CSBEL011628DO
Dog Interleukin-1 receptor antagonist protein (IL1RN) ELISA kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma tissue homogenates Method Sandwich