Learn More
Biomatik Corporation Dog Interleukin 12A (IL12A) ELISA Kit
Supplier:  Biomatik Corporation EKU05197
Encompass_Preferred
Dog Interleukin 12A (IL12A) ELISA Kit for detecting IL12A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q28267. Target Synonyms: p35; IL12-A; CLMF1; NFSK; NKSF1; Natural Killer Cell Stimulatory Factor 1; Cytotoxic Lymphocyte Maturation Factor 1; NF Cell Stimulatory Factor Chain 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6591