Learn More
Cusabio Technology LLC Dog Interleukin-33 IL33 ELISA
Catalog No. 5000049753
Encompass_Preferred
This kit detects IL33 in serum plasma and tissue homogenates with a quantitative sandwich ELISA method It offers a detection range of 23 44 pg/mL to 1500 pg/mL and a sensitivity of 5 86 pg/mL ensuring precise measurement of canine interleukin-33 levels The assay requires 50-100 L of sample per test and delivers results within 1-5 hours using a 450 nm wavelength for detection Designed for reliability it is suitable for researchers studying immune responses or inflammatory conditions in dogs providing robust data for both experimental and diagnostic applications
Catalog No. 50-000-49753 Supplier Cusabio Technology LLC Supplier No. CSBEL011656DO
This kit detects IL33 in serum plasma and tissue homogenates with a quantitative sandwich ELISA method It offers a detection range of 23 44 pg/mL to 1500 pg/mL and a sensitivity of 5 86 pg/mL ensuring precise measurement of canine interleukin-33 levels The assay requires 50-100 L of sample per test and delivers results within 1-5 hours using a 450 nm wavelength for detection Designed for reliability it is suitable for researchers studying immune responses or inflammatory conditions in dogs providing robust data for both experimental and diagnostic applications