Learn More
American Research Products Inc Dog Interleukin 6 IL 6 ELISA
Catalog No. 5000329639
Encompass
Dog Interleukin 6 IL-6 ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29639 Supplier American Research Products Inc Supplier No. CSBE11260C
Dog Interleukin 6 IL-6 ELISA Kit from CUSABIO Detection Range 1 56 pg/mL-100 pg/mL Sensitivity 0 39 pg/mL Sample serum plasma tissue homogenates Method Sandwich