Learn More
American Research Products Inc Dog Interleukin 8 IL 8 ELISA
Catalog No. 5000328803
Encompass
Dog Interleukin-8 IL-8 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 146 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich
Catalog No. 50-003-28803 Supplier American Research Products Inc Supplier No. CSBE08052C
Dog Interleukin-8 IL-8 ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 146 ng/mL Sample serum plasma cell culture supernates tissue homogenates Method Sandwich