Learn More
Cayman Chemical Dog LIvR X ReceptrBeta Re 96
SDP
Catalog No. 502942824
Encompass_Preferred
INDIGO Bioscience’s assays are all-inclusive cell-based assay systems. In addition to LXRβ Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist TO901317, Luciferase Detection Reagent, and a cell culture-ready assay plate
Catalog No. 50-294-2824 Supplier Cayman Chemical Supplier No. 4103196 WELL
INDIGO Bioscience’s assays are all-inclusive cell-based assay systems. In addition to LXRβ Reporter Cells, this kit provides two optimized media for use during cell culture and in diluting the user's test samples, the reference agonist TO901317, Luciferase Detection Reagent, and a cell culture-ready assay plate