Learn More
American Research Products Inc Dog Osteocalcin bone gla prote
Catalog No. 5000329009
Encompass
Dog Osteocalcin/bone gla protein OT/BGP ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29009 Supplier American Research Products Inc Supplier No. CSBE13345C
Dog Osteocalcin/bone gla protein OT/BGP ELISA Kit from CUSABIO Detection Range 0 312 ng/mL-20 ng/mL Sensitivity 0 078 ng/mL Sample serum plasma tissue homogenates Method Sandwich