Learn More
Biomatik Corporation Dog Osteocalcin (OC) ELISA Kit
Supplier:  Biomatik Corporation EKU09001
Encompass_Preferred
Dog Osteocalcin (OC) ELISA Kit for detecting OC in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 0.625-40ng/ml. Sensitivity: 0.249ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P81455. Target Synonyms: BGLAP; OT; BGP; Bone Gla Protein; Bone Gamma-Carboxyglutamate Protein; Gamma-carboxyglutamic acid-containing protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6341