Learn More
Cayman Chemical Dog Pregnane X Receptor Reporter Assay System, 3x32 Well
SDP
Catalog No. 502964360
Encompass_Preferred
dPXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.[INDIGO Catalog Nos. D07001-32, D07001]
Catalog No. 50-296-4360 Supplier Cayman Chemical Supplier No. 214023X32 WELL
dPXR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields exceptional cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for cumbersome intermediate treatment steps such as spin-and-rinse of cells, viability determinations, or cell titer adjustments prior to assay setup.[INDIGO Catalog Nos. D07001-32, D07001]