Learn More
Biomatik Corporation Dog Procalcitonin (PCT) ELISA Kit
Supplier:  Biomatik Corporation EKU12425
Encompass_Preferred
Dog Procalcitonin (PCT) ELISA Kit for detecting PCT in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41547. Target Synonyms: Pro-Calcitonin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7586