Learn More
Cayman Chemical Dog PRoxIsome ProlIfRatrA 96
SDP
Catalog No. 502939871
Encompass_Preferred
dPPARδ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. D00121-32, D00121]
Catalog No. 50-293-9871 Supplier Cayman Chemical Supplier No. 2831396 WELL
dPPARδ Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments.[INDIGO Catalog Nos. D00121-32, D00121]