Learn More
Biomatik Corporation Dog Substance P (SP) CLIA Kit
Supplier:  Biomatik Corporation EKU08613
Encompass_Preferred
Dog Substance P (SP) CLIA Kit for detecting SP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 11.72-3,000pg/ml. Sensitivity: 4.85pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: E2RF69. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4963