Learn More
American Research Products Inc Dog Transforming growth factor
Catalog No. 5000329090
Encompass
Dog Transforming growth factor b1 TGF-b1 ELISA Kit from CUSABIO Detection Range 0 781 ng/ml - 50 ng/ml Sensitivity 0 747 ng/ml Sample serum plasma Method Sandwich
Catalog No. 50-003-29090 Supplier American Research Products Inc Supplier No. CSBE04796C
Dog Transforming growth factor b1 TGF-b1 ELISA Kit from CUSABIO Detection Range 0 781 ng/ml - 50 ng/ml Sensitivity 0 747 ng/ml Sample serum plasma Method Sandwich