Learn More
Biomatik Corporation Dog Tryptase (TPS) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10968
Encompass_Preferred
Dog Tryptase (TPS) High-sensitive ELISA Kit for detecting TPS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P15944. Gene ID: 100049001. Target Synonyms: TPSAB1; Tryptase Alpha/Beta 1; Tryptase Alpha II; Tryptase Beta I; Tryptase-I; Tryptase-II; Tryptase-III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8260