Learn More
Crystal Chem Inc Dog TSH ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80566
Encompass
The Dog Thyroid Stimulating Hormone of TSH) ELISA is a highly sensitive assay that is used to quantify levels of canine TSH in serum and plasma. Highlights: Kit uses sample size of only 100 µL. Dynamic range of 2.5 - 80 ng/mL. Highly precise with low CV's.
Catalog No. 50-194-7734