Learn More
American Research Products Inc Dog Tumor necrosis factor a T
Catalog No. 5000329771
Encompass
Dog Tumor necrosis factor a TNF-a ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29771 Supplier American Research Products Inc Supplier No. CSBE11737C
Dog Tumor necrosis factor a TNF-a ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates Method Sandwich